Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145950.4 | NP_666062.3 | 4 - 5 | 672 | 124 | |
| GenBank mRNA | AK048731.1 | - | 4 - 5 | 673 | 124 | |
| AK148243.1 | - | 4 - 5 | 647 | 124 | ||
| AK169678.1 | - | 4 - 5 | 617 | 124 | ||
| CB574822.1 | - | 4 - 5 | 628 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osgin2
Genomic Map