Assay Details
| Gene Symbol: Cabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160252.1 | NP_001153724.1 | 5 - 6 | 527 | 58 | |
| NM_001160253.1 | NP_001153725.1 | 5 - 6 | 809 | 58 | ||
| NM_013878.2 | NP_038906.2 | 6 - 7 | 686 | 58 | ||
| GenBank mRNA | AF169156.1 | - | 5 - 6 | 555 | 58 | |
| AF169157.1 | - | 6 - 7 | 714 | 58 | ||
| AK007488.1 | - | 5 - 6 | 809 | 58 | ||
| BC107029.2 | - | 6 - 7 | 686 | 40050356 | 58 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | AF169156.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | AF169157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | AK007488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | BC107029.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | NM_001160252.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | AF169156.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | AF169157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | AK007488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | BC107029.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187625 | NM_001160252.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631428 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | AF169156.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | AF169157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | AK007488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | BC107029.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | NM_001160252.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | AF169156.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | AF169157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | AK007488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | BC107029.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187625 | NM_001160252.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920960 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03540556 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09089622 | - | - |
| Whole Mouse Genome G4122A | A_51_P187625 | AF169156.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | AF169157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | AK007488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | BC107029.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | NM_001160252.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | AF169156.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | AF169157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | AK007488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | BC107029.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187625 | NM_001160252.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107987 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109781 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14473029 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14473009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14473006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14473018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14473019 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14473008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14473008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14324742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14221500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14221502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14472996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14141771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14473016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14472999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460311 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460306 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460308 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420597_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460303 | - | - |
| Mouse Genome 430 2.0 Array | 1420597_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420597_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 29866_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 29866_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | NM_001160252.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | NM_001160252.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | NM_001160253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | NM_001160253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | NM_013878.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | NM_013878.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000315.mm.1 | TC1900000025.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | NM_001160252.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | NM_001160252.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | NM_001160253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | NM_001160253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | NM_013878.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | NM_013878.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000316.mm.1 | TC1900000025.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | NM_001160252.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | NM_001160252.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | NM_001160253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | NM_001160253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | NM_013878.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | NM_013878.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000314.mm.1 | TC1900000025.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420597_PM_a_at | - | - |
| Mouse Expression Set 430 | 1420597_a_at | - | - |
| Murine Genome U74v2 Set | 166983_at | - | - |
| Murine Genome U74v2 Set | 166983_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420597_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420597_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420597_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420597_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 914630 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 840142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000840142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2713868 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2713868 | - | - |
| mouse-6 v1.1 (Illumina) | scl52800.8.1_155-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 840142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52800.8.1_155-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2915898 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2915898 | - | - |

Cabp2
Genomic Map