Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 12 - 13 | 1418 | 64 | |
| GenBank mRNA | AK084205.1 | - | 12 - 13 | 1365 | 64 | |
| AK146353.1 | - | 12 - 13 | 1382 | 64 | ||
| AK149921.1 | - | 12 - 13 | 1365 | 64 | ||
| AK154918.1 | - | 12 - 13 | 1365 | 64 | ||
| AK173267.1 | - | 12 - 13 | 1393 | 64 | ||
| BC021520.1 | - | 10 - 11 | 1212 | 64 | ||
| BC066136.1 | - | 11 - 12 | 1171 | 64 | ||
| BC089160.1 | - | 12 - 13 | 1340 | 5390706 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp512
Genomic Map