Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 4 - 5 | 492 | 58 | |
| GenBank mRNA | AK146353.1 | - | 4 - 5 | 456 | 58 | |
| AK149921.1 | - | 4 - 5 | 439 | 58 | ||
| AK154918.1 | - | 4 - 5 | 439 | 58 | ||
| AK173267.1 | - | 4 - 5 | 467 | 58 | ||
| BC021520.1 | - | 2 - 3 | 302 | 58 | ||
| BC066136.1 | - | 4 - 5 | 415 | 58 | ||
| BC089160.1 | - | 4 - 5 | 414 | 5390706 | 58 | |
More Information
Set Membership: |
|

Zfp512
Genomic Map