Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 7 - 8 | 792 | 77 | |
| GenBank mRNA | AK084205.1 | - | 7 - 8 | 739 | 77 | |
| AK146353.1 | - | 7 - 8 | 756 | 77 | ||
| AK149921.1 | - | 7 - 8 | 739 | 77 | ||
| AK154918.1 | - | 7 - 8 | 739 | 77 | ||
| AK173267.1 | - | 7 - 8 | 767 | 77 | ||
| BC021520.1 | - | 5 - 6 | 602 | 77 | ||
| BC066136.1 | - | 7 - 8 | 715 | 77 | ||
| BC089160.1 | - | 7 - 8 | 714 | 5390706 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp512
Genomic Map