Assay Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173764.3 | NP_776125.2 | 2 - 3 | 319 | 84 | |
| GenBank mRNA | BC110670.1 | - | 1 - 2 | 40 | 84 | |
| CN459607.1 | - | 2 - 3 | 319 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tapt1
Genomic Map