Assay Details
| Gene Symbol: Smim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025471.2 | NP_079747.1 | 1 - 2 | 115 | 71 | |
| GenBank mRNA | AK005941.1 | - | 1 - 2 | 118 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005941.1, NM_025471.2 transcripts.
Set Membership: |
|

Smim8
Genomic Map