Assay Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016890.4 | NP_058586.3 | 5 - 6 | 444 | 55 | |
| GenBank mRNA | AK002720.1 | - | 5 - 6 | 435 | 55 | |
| AK003255.1 | - | 5 - 6 | 419 | 55 | ||
| AK011840.1 | - | 5 - 6 | 441 | 55 | ||
| AK159344.1 | - | 5 - 6 | 442 | 55 | ||
| AK182271.1 | - | 1 - 2 | 39 | 55 | ||
| BC061120.1 | - | 5 - 6 | 425 | 6774742 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dctn3
Genomic Map