Assay Details
| Gene Symbol: Prdm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080771.1 | NP_001074240.1 | 2 - 3 | 586 | 67 | |
| GenBank mRNA | AK149252.1 | - | 2 - 3 | 469 | 67 | |
| BB665287.1 | - | 2 - 3 | 406 | 67 | ||
| BC017632.1 | - | 2 - 3 | 586 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdm13
Genomic Map