Assay Details
| Gene Symbol: Dpy19l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081201.1 | NP_001074670.1 | 8 - 9 | 976 | 128 | |
| GenBank mRNA | AK144417.1 | - | 8 - 9 | 928 | 128 | |
| BC147500.1 | - | 8 - 9 | 932 | 9056363 | 128 | |
| BC157932.1 | - | 8 - 9 | 932 | 9088105 | 128 | |
| BC157988.1 | - | 8 - 9 | 932 | 9088143 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l4
Genomic Map