Assay Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145558.2 | NP_663533.1 | 1 - 2 | 81 | 98 | |
| GenBank mRNA | AK076814.1 | - | 1 - 2 | 44 | 98 | |
| AK083767.1 | - | 1 - 2 | 44 | 98 | ||
| AK150889.1 | - | 1 - 2 | 43 | 98 | ||
| AK168594.1 | - | 1 - 2 | 45 | 98 | ||
| AK171803.1 | - | 1 - 2 | 46 | 98 | ||
| BY337615.1 | - | 1 - 2 | 82 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK076814.1,AK083767.1,AK150889.1,NM_145558.2 transcripts.
This assay is in the 5'UTR region of the AK076814.1,AK083767.1,AK150889.1,NM_145558.2 transcripts.
Set Membership: |
|

Hadhb
Genomic Map