Assay Details
| Gene Symbol: Mtus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002529.1 | - | 1 - 2 | 178 | 82 | |
| AK021162.1 | - | 1 - 2 | 209 | 82 | ||
| BC055016.1 | - | 1 - 2 | 249 | 82 | ||
More Information
Set Membership: |
|

Mtus2
Genomic Map