Assay Details
| Gene Symbol: Mtus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029920.7 | NP_084196.4 | 8 - 9 | 4073 | 70 | |
| GenBank mRNA | AB093263.1 | - | 7 - 8 | 2948 | 70 | |
| AK002529.1 | - | 2 - 3 | 245 | 70 | ||
| AK021162.1 | - | 2 - 3 | 276 | 70 | ||
| AK147457.1 | - | 8 - 9 | 4074 | 70 | ||
| BC055016.1 | - | 2 - 3 | 316 | 70 | ||
| BC085194.1 | - | 8 - 9 | 3419 | 70 | ||
| BC150942.1 | - | 8 - 9 | 3507 | 9087853 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtus2
Genomic Map