Assay Details
| Gene Symbol: Mtus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029920.7 | NP_084196.4 | 9 - 10 | 4288 | 125 | |
| GenBank mRNA | AK002529.1 | - | 3 - 4 | 460 | 125 | |
| AK147457.1 | - | 9 - 10 | 4289 | 125 | ||
| BC055016.1 | - | 3 - 4 | 531 | 125 | ||
| BC085194.1 | - | 9 - 10 | 3634 | 125 | ||
| BC150942.1 | - | 9 - 10 | 3722 | 9087853 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtus2
Genomic Map