Assay Details
| Gene Symbol: Mtus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029920.7 | NP_084196.4 | 12 - 13 | 4567 | 100 | |
| GenBank mRNA | AB093263.1 | - | 11 - 12 | 3441 | 100 | |
| AK021162.1 | - | 6 - 7 | 769 | 100 | ||
| AK147457.1 | - | 12 - 13 | 4568 | 100 | ||
| BC055016.1 | - | 6 - 7 | 810 | 100 | ||
| BC085194.1 | - | 12 - 13 | 3913 | 100 | ||
More Information
Set Membership: |
|

Mtus2
Genomic Map