Assay Details
| Gene Symbol: Cpsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172302.3 | NP_758506.3 | 5 - 6 | 693 | 90 | |
| GenBank mRNA | AK044306.1 | - | 5 - 6 | 673 | 90 | |
| AK053375.1 | - | 6 - 7 | 738 | 90 | ||
| AK088603.1 | - | 5 - 6 | 671 | 90 | ||
| AK165330.1 | - | 5 - 6 | 671 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK053375.1 transcripts.
Set Membership: |
|

Cpsf7
Genomic Map