Assay Details
| Gene Symbol: Rgs19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291207.1 | NP_001278136.1 | 1 - 2 | 102 | 65 | |
| GenBank mRNA | AY138505.1 | - | 1 - 2 | 54 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY138505.1, NM_001291207.1 transcripts.
Set Membership: |
|

Rgs19
Genomic Map