Assay Details
| Gene Symbol: Sim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011377.2 | NP_035507.2 | 9 - 10 | 1727 | 69 | |
| GenBank mRNA | D63383.1 | - | 9 - 10 | 1733 | 69 | |
| D64135.1 | - | 9 - 10 | 1674 | 69 | ||
| U42554.1 | - | 9 - 10 | 1399 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sim2
Genomic Map