Assay Details
| Gene Symbol: S100a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163525.2 | NP_001156997.1 | 3 - 4 | 337 | 83 | |
| NM_025393.4 | NP_079669.2 | 3 - 4 | 320 | 83 | ||
| GenBank mRNA | AA726094.1 | - | 3 - 4 | 330 | 83 | |
| AK003669.1 | - | 3 - 4 | 300 | 83 | ||
| AK019075.1 | - | 3 - 4 | 291 | 83 | ||
| BC025607.1 | - | 2 - 3 | 319 | 5151924 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

S100a14
Genomic Map