Assay Details
| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010252.4 | NP_034382.2 | 4 - 5 | 940 | 98 | |
| GenBank mRNA | AK032128.1 | - | 4 - 5 | 625 | 98 | |
| AK034085.1 | - | 4 - 5 | 528 | 98 | ||
| AK034332.1 | - | 4 - 5 | 941 | 98 | ||
| AK038468.1 | - | 4 - 5 | 677 | 98 | ||
| AK039068.1 | - | 4 - 5 | 638 | 98 | ||
| AK047752.1 | - | 4 - 5 | 690 | 98 | ||
| AK049956.1 | - | 4 - 5 | 678 | 98 | ||
| AK140399.1 | - | 4 - 5 | 690 | 98 | ||
| AK162884.1 | - | 5 - 6 | 623 | 98 | ||
| AK163541.1 | - | 4 - 5 | 648 | 98 | ||
| BC099939.1 | - | 4 - 5 | 649 | 30090762 | 98 | |
| X55272.1 | - | 4 - 5 | 723 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabrg1
Genomic Map