Assay Details
| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010252.4 | NP_034382.2 | 5 - 6 | 1017 | 128 | |
| GenBank mRNA | AK032128.1 | - | 5 - 6 | 702 | 128 | |
| AK034085.1 | - | 5 - 6 | 605 | 128 | ||
| AK034332.1 | - | 5 - 6 | 1018 | 128 | ||
| AK038468.1 | - | 5 - 6 | 754 | 128 | ||
| AK039068.1 | - | 5 - 6 | 715 | 128 | ||
| AK047752.1 | - | 5 - 6 | 767 | 128 | ||
| AK049956.1 | - | 5 - 6 | 755 | 128 | ||
| AK140399.1 | - | 5 - 6 | 767 | 128 | ||
| AK162884.1 | - | 6 - 7 | 700 | 128 | ||
| AK163541.1 | - | 5 - 6 | 725 | 128 | ||
| BC099939.1 | - | 5 - 6 | 726 | 30090762 | 128 | |
| X55272.1 | - | 5 - 6 | 800 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabrg1
Genomic Map