Assay Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024434.6 | NP_077754.3 | 11 - 12 | 1348 | 56 | |
| GenBank mRNA | AF334160.1 | - | 11 - 12 | 1309 | 56 | |
| AK002819.1 | - | 11 - 12 | 1284 | 56 | ||
| AK005334.1 | - | 11 - 12 | 1261 | 56 | ||
| AK008600.1 | - | 11 - 12 | 1235 | 56 | ||
| AK010384.1 | - | 11 - 12 | 1232 | 56 | ||
| AK010502.1 | - | 11 - 12 | 1301 | 56 | ||
| AK011604.1 | - | 11 - 12 | 1234 | 56 | ||
| AK166958.1 | - | 11 - 12 | 1232 | 56 | ||
| AK169032.1 | - | 11 - 12 | 1234 | 56 | ||
| AK185578.1 | - | 3 - 4 | 245 | 56 | ||
| BC016536.1 | - | 11 - 12 | 1348 | 3488469 | 56 | |
| CT010237.1 | - | 11 - 12 | 1262 | 56 | ||
| CT010301.1 | - | 11 - 12 | 1262 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lap3
Genomic Map