Assay Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024434.6 | NP_077754.3 | 6 - 7 | 792 | 80 | |
| GenBank mRNA | AF334160.1 | - | 6 - 7 | 753 | 80 | |
| AK002819.1 | - | 6 - 7 | 728 | 80 | ||
| AK005334.1 | - | 6 - 7 | 705 | 80 | ||
| AK008600.1 | - | 6 - 7 | 679 | 80 | ||
| AK010384.1 | - | 6 - 7 | 676 | 80 | ||
| AK010502.1 | - | 6 - 7 | 745 | 80 | ||
| AK011604.1 | - | 6 - 7 | 678 | 80 | ||
| AK089023.1 | - | 6 - 7 | 759 | 80 | ||
| AK166958.1 | - | 6 - 7 | 676 | 80 | ||
| AK169032.1 | - | 6 - 7 | 678 | 80 | ||
| AK186195.1 | - | 1 - 2 | 150 | 80 | ||
| BC016536.1 | - | 6 - 7 | 792 | 3488469 | 80 | |
| CT010237.1 | - | 6 - 7 | 706 | 80 | ||
| CT010301.1 | - | 6 - 7 | 706 | 80 | ||
More Information
Set Membership: |
|

Lap3
Genomic Map