Assay Details
| Gene Symbol: Fam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020622.2 | NP_065647.1 | 1 - 2 | 115 | 92 | |
| GenBank mRNA | AF360358.1 | - | 1 - 2 | 116 | 92 | |
| AK018571.1 | - | 1 - 2 | 85 | 92 | ||
| BC048949.1 | - | 1 - 2 | 62 | 6474753 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF360358.1, AK018571.1, BC048949.1, NM_020622.2 transcripts.
Set Membership: |
|

Fam3b
Genomic Map