Assay Details
| Gene Symbol: Fam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020622.2 | NP_065647.1 | 3 - 4 | 377 | 66 | |
| GenBank mRNA | AF360358.1 | - | 3 - 4 | 378 | 66 | |
| AF494380.1 | - | 3 - 4 | 282 | 66 | ||
| AK018571.1 | - | 3 - 4 | 347 | 66 | ||
| BC048949.1 | - | 3 - 4 | 324 | 6474753 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam3b
Genomic Map