| Array Name |
Probe IDs |
Targets |
Target Type |
| Mouse Genome 430 2.0 Array |
1434569_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1434569_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500020746.mm.1 |
NM_001170454.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020746.mm.1 |
NM_001170454.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020746.mm.1 |
TC0500002258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020747.mm.1 |
NM_001170454.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020747.mm.1 |
NM_001170454.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020747.mm.1 |
TC0500002258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020748.mm.1 |
NM_001170454.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020748.mm.1 |
NM_001170454.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020748.mm.1 |
TC0500002258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020750.mm.1 |
NM_001170454.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020750.mm.1 |
NM_001170454.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020750.mm.1 |
TC0500002258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020749.mm.1 |
NM_001170454.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020749.mm.1 |
NM_001170454.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020749.mm.1 |
TC0500002258.mm.1 |
MTA TC |
| Murine Genome U74v2 Set |
134206_f_at |
- |
- |
| Murine Genome U74v2 Set |
134206_PM_f_at |
- |
- |