Assay Details
| Gene Symbol: Pabpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053114.2 | NP_444344.1 | 1 - 2 | 110 | 70 | |
| GenBank mRNA | AK050551.1 | - | 1 - 2 | 112 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050551.1, NM_053114.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625282 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625284 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625283 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040588 | BC107363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040588 | NM_053114.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040588 | AK050551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040588 | BC107362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040594 | NM_053114.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040594 | AK050551.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659133 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139416 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040594 | NM_053114.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040594 | AK050551.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040588 | BC107363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040588 | NM_053114.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040588 | AK050551.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040588 | BC107362.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440187 | NM_053114.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440187 | AK050551.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32625292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139416 | - | - |
| Whole Mouse Genome G4122A | A_51_P440187 | NM_053114.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440187 | AK050551.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106553 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14918080 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14890410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14890409 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14890406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14918084 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625282 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32625284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32625345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14890411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14918092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15151516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32625309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601564 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601562 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1446162_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601561 | - | - |
| Mouse Genome 430 2.0 Array | 1446162_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446162_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93728_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93728_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00007508.mm.1 | NM_053114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007508.mm.1 | NM_053114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007508.mm.1 | TC0X00001172.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00007509.mm.1 | NM_053114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007509.mm.1 | NM_053114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007509.mm.1 | TC0X00001172.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00007514.mm.1 | NM_053114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007514.mm.1 | NM_053114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007514.mm.1 | TC0X00001172.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00007512.mm.1 | NM_053114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007512.mm.1 | NM_053114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007512.mm.1 | TC0X00001172.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00007513.mm.1 | NM_053114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007513.mm.1 | NM_053114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007513.mm.1 | TC0X00001172.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1446162_at | - | - |
| Mouse Expression Set 430 | 1446162_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446162_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446162_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 350892 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2622755 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2622755 | - | - |
| mouse-6 v1.1 (Illumina) | scl53886.2.1_262-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53886.2.1_262-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2622755 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2622755 | - | - |

Pabpc5
Genomic Map