| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900011219.mm.1 |
NM_010730.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011219.mm.1 |
NM_010730.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011219.mm.1 |
TC1900001240.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011222.mm.1 |
NM_010730.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011222.mm.1 |
NM_010730.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011222.mm.1 |
TC1900001240.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011221.mm.1 |
NM_010730.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011221.mm.1 |
NM_010730.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011221.mm.1 |
TC1900001240.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011216.mm.1 |
NM_010730.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011216.mm.1 |
NM_010730.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011216.mm.1 |
TC1900001240.mm.1 |
MTA TC |