Assay Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159565.1 | NP_001153037.1 | 3 - 4 | 300 | 59 | |
| NM_016890.4 | NP_058586.3 | 3 - 4 | 300 | 59 | ||
| GenBank mRNA | AF098508.1 | - | 3 - 4 | 279 | 59 | |
| AK002720.1 | - | 3 - 4 | 291 | 59 | ||
| AK003255.1 | - | 3 - 4 | 275 | 59 | ||
| AK011840.1 | - | 3 - 4 | 297 | 59 | ||
| AK159344.1 | - | 3 - 4 | 298 | 59 | ||
| AV493721.1 | - | 3 - 4 | 286 | 59 | ||
| BC061120.1 | - | 3 - 4 | 281 | 6774742 | 59 | |
| BY141790.1 | - | 3 - 4 | 307 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dctn3
Genomic Map