Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290700.1 | NP_001277629.1 | 3 - 4 | 421 | 69 | |
| GenBank mRNA | AK015797.1 | - | 3 - 4 | 396 | 69 | |
| AK122541.1 | - | 3 - 4 | 246 | 69 | ||
| AK210231.1 | - | 2 - 3 | 168 | 69 | ||
| BC019546.1 | - | 3 - 4 | 257 | 4038791 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp451
Genomic Map