Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290699.1 | NP_001277628.1 | 9 - 10 | 2738 | 120 | |
| NM_133817.3 | NP_598578.1 | 11 - 12 | 2976 | 120 | ||
| GenBank mRNA | AK030088.1 | - | 11 - 12 | 2809 | 120 | |
| BC024435.1 | - | 2 - 3 | 651 | 120 | ||
| BC026587.1 | - | 2 - 3 | 1154 | 120 | ||
| BC043685.1 | - | 2 - 3 | 1226 | 120 | ||
| BC062154.1 | - | 9 - 10 | 2528 | 6331597 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp451
Genomic Map