Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290699.1 | NP_001277628.1 | 10 - 11 | 2869 | 65 | |
| NM_133817.3 | NP_598578.1 | 12 - 13 | 3107 | 65 | ||
| GenBank mRNA | AK030088.1 | - | 12 - 13 | 2940 | 65 | |
| BC024435.1 | - | 3 - 4 | 782 | 65 | ||
| BC026587.1 | - | 3 - 4 | 1285 | 65 | ||
| BC043685.1 | - | 3 - 4 | 1357 | 65 | ||
| BC062154.1 | - | 10 - 11 | 2659 | 6331597 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp451
Genomic Map