Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133817.3 | NP_598578.1 | 4 - 5 | 542 | 86 | |
| GenBank mRNA | AK030088.1 | - | 4 - 5 | 375 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp451
Genomic Map