Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290699.1 | NP_001277628.1 | 7 - 8 | 1003 | 61 | |
| NM_133817.3 | NP_598578.1 | 9 - 10 | 1241 | 61 | ||
| GenBank mRNA | AK030088.1 | - | 9 - 10 | 1074 | 61 | |
| BC062154.1 | - | 7 - 8 | 793 | 6331597 | 61 | |
More Information
Set Membership: |
|

Zfp451
Genomic Map