Assay Details
| Gene Symbol: Yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207633.2 | NP_997516.1 | 1 - 2 | 504 | 131 | |
| GenBank mRNA | AK020780.1 | - | 1 - 2 | 78 | 131 | |
| AK030473.1 | - | 1 - 2 | 506 | 131 | ||
| AK045453.1 | - | 1 - 2 | 216 | 131 | ||
| AK053562.1 | - | 1 - 2 | 505 | 131 | ||
| AK079683.1 | - | 1 - 2 | 116 | 131 | ||
| BB642656.1 | - | 1 - 2 | 525 | 131 | ||
| BC048712.1 | - | 1 - 2 | 139 | 6328657 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045453.1, AK079683.1, BC048712.1, NM_207633.2 transcripts.
Set Membership: |
|

Yipf6
Genomic Map