Assay Details
| Gene Symbol: Yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207633.2 | NP_997516.1 | 3 - 4 | 716 | 110 | |
| GenBank mRNA | AK020780.1 | - | 3 - 4 | 290 | 110 | |
| AK030473.1 | - | 3 - 4 | 718 | 110 | ||
| AK045453.1 | - | 3 - 4 | 428 | 110 | ||
| AK053562.1 | - | 3 - 4 | 717 | 110 | ||
| AK079683.1 | - | 3 - 4 | 328 | 110 | ||
| AK143152.1 | - | 3 - 4 | 286 | 110 | ||
| AK160251.1 | - | 4 - 5 | 344 | 110 | ||
| BC048712.1 | - | 3 - 4 | 351 | 6328657 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yipf6
Genomic Map