Assay Details
| Gene Symbol: Yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207633.2 | NP_997516.1 | 5 - 6 | 887 | 75 | |
| GenBank mRNA | AK020780.1 | - | 5 - 6 | 76 | ||
| AK030473.1 | - | 5 - 6 | 889 | 75 | ||
| AK045453.1 | - | 5 - 6 | 599 | 75 | ||
| AK053562.1 | - | 5 - 6 | 74 | |||
| AK079683.1 | - | 5 - 6 | 499 | 75 | ||
| AK143152.1 | - | 5 - 6 | 457 | 75 | ||
| AK160251.1 | - | 6 - 7 | 515 | 75 | ||
| BC048712.1 | - | 5 - 6 | 522 | 6328657 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yipf6
Genomic Map