Assay Details
Gene Symbol: Atp6v1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133826.4 | NP_598587.2 | 2 - 3 | 256 | 111 | |
GenBank mRNA | AK078767.1 | - | 2 - 3 | 257 | 111 | |
AK081492.1 | - | 2 - 3 | 266 | 111 | ||
AK151659.1 | - | 2 - 3 | 319 | 111 | ||
AK152147.1 | - | 2 - 3 | 277 | 111 | ||
BC009154.1 | - | 2 - 3 | 257 | 3601621 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
