Assay Details
Gene Symbol: Atp6v1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133826.4 | NP_598587.2 | 7 - 8 | 715 | 71 | |
GenBank mRNA | AK078767.1 | - | 7 - 8 | 716 | 71 | |
AK152147.1 | - | 7 - 8 | 736 | 71 | ||
BC009154.1 | - | 7 - 8 | 716 | 3601621 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
