Assay Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277101.1 | NP_001264030.1 | 2 - 3 | 477 | 83 | |
| NM_013715.2 | NP_038743.1 | 2 - 3 | 726 | 83 | ||
| NR_102282.1 | 2 - 3 | 734 | 83 | |||
| GenBank mRNA | AF065386.1 | - | 2 - 3 | 479 | 83 | |
| AF068223.1 | - | 2 - 3 | 494 | 83 | ||
| AK002761.1 | - | 2 - 3 | 508 | 83 | ||
| AK005728.1 | - | 2 - 3 | 513 | 83 | ||
| AK012499.1 | - | 2 - 3 | 83 | |||
| AK133073.1 | - | 2 - 3 | 506 | 83 | ||
| AK146271.1 | - | 2 - 3 | 513 | 83 | ||
| AK151492.1 | - | 2 - 3 | 512 | 83 | ||
| BC046753.1 | - | 2 - 3 | 472 | 5693100 | 83 | |
| CX240457.1 | - | 2 - 3 | 694 | 83 | ||
| DV056728.1 | - | 2 - 3 | 533 | 83 | ||
| U70736.1 | - | 2 - 3 | 502 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cops5
Genomic Map