Assay Details
| Gene Symbol: Arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033329.3 | NP_001028501.1 | 5 - 6 | 1051 | 81 | |
| NM_001290384.1 | NP_001277313.1 | 4 - 5 | 673 | 81 | ||
| NM_001290385.1 | NP_001277314.1 | 4 - 5 | 1497 | 81 | ||
| GenBank mRNA | AK038840.1 | - | 5 - 6 | 969 | 81 | |
| AK122280.1 | - | 4 - 5 | 608 | 81 | ||
| AK139416.1 | - | 5 - 6 | 999 | 81 | ||
| AK163874.1 | - | 4 - 5 | 654 | 81 | ||
| BC067031.1 | - | 4 - 5 | 1319 | 81 | ||
| BC141385.1 | - | 4 - 5 | 693 | 9055998 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef9
Genomic Map