Assay Details
| Gene Symbol: Arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033329.3 | NP_001028501.1 | 6 - 7 | 1276 | 65 | |
| NM_001290384.1 | NP_001277313.1 | 5 - 6 | 898 | 65 | ||
| NM_001290385.1 | NP_001277314.1 | 5 - 6 | 1722 | 65 | ||
| GenBank mRNA | AK038840.1 | - | 6 - 7 | 1194 | 65 | |
| AK122280.1 | - | 5 - 6 | 833 | 65 | ||
| AK139416.1 | - | 6 - 7 | 1224 | 65 | ||
| AK163874.1 | - | 5 - 6 | 879 | 65 | ||
| BC067031.1 | - | 5 - 6 | 1544 | 65 | ||
| BC141385.1 | - | 5 - 6 | 918 | 9055998 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef9
Genomic Map