Assay Details
| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010250.5 | NP_034380.1 | 5 - 6 | 1259 | 83 | |
| GenBank mRNA | AK034095.1 | - | 5 - 6 | 1260 | 83 | |
| AK039496.1 | - | 5 - 6 | 543 | 83 | ||
| BC132331.1 | - | 5 - 6 | 935 | 40130608 | 83 | |
| BC137733.1 | - | 5 - 6 | 938 | 8860752 | 83 | |
| M63436.1 | - | 5 - 6 | 932 | 83 | ||
| M86566.1 | - | 5 - 6 | 627 | 83 | ||
| X61430.1 | - | 4 - 5 | 515 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabra1
Genomic Map