Assay Details
| Gene Symbol: Hcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081218.1 | NP_001074687.1 | 9 - 10 | 1415 | 114 | |
| GenBank mRNA | BC116411.1 | - | 2 - 3 | 207 | 114 | |
| BC127176.1 | - | 2 - 3 | 207 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hcfc2
Genomic Map