| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
NM_001146690.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
NM_001146690.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
NM_144807.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
NM_144807.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
NR_027477.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
NR_027477.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020007.mm.1 |
TC1000002645.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
NM_001146690.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
NM_001146690.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
NM_144807.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
NM_144807.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
NR_027477.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
NR_027477.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020005.mm.1 |
TC1000002645.mm.1 |
MTA TC |