Assay Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114361.1 | NP_001107833.1 | 12 - 13 | 1532 | 84 | |
| NM_001114362.1 | NP_001107834.1 | 10 - 11 | 1325 | 84 | ||
| NM_001286567.1 | NP_001273496.1 | 12 - 13 | 1435 | 84 | ||
| NM_199466.3 | NP_955760.3 | 9 - 10 | 1196 | 84 | ||
| GenBank mRNA | AK077527.1 | - | 4 - 5 | 359 | 84 | |
| AK144604.1 | - | 12 - 13 | 1533 | 84 | ||
| BC065178.1 | - | 12 - 13 | 1373 | 6827397 | 84 | |
| BC066099.1 | - | 9 - 10 | 1127 | 30533573 | 84 | |
| BC067011.1 | - | 9 - 10 | 1145 | 6405184 | 84 | |
| BC070427.1 | - | 10 - 11 | 1265 | 6413041 | 84 | |
| BC079619.1 | - | 9 - 10 | 1127 | 30533573 | 84 | |
| DQ230326.1 | - | 11 - 12 | 1215 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eml4
Genomic Map