| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100000389.mm.1 |
NM_177722.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000389.mm.1 |
NM_177722.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000389.mm.1 |
TC0100000045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000396.mm.1 |
NM_177722.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000396.mm.1 |
NM_177722.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000396.mm.1 |
TC0100000045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000395.mm.1 |
NM_177722.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000395.mm.1 |
NM_177722.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000395.mm.1 |
TC0100000045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000398.mm.1 |
NM_177722.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000398.mm.1 |
NM_177722.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000398.mm.1 |
TC0100000045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000397.mm.1 |
NM_177722.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000397.mm.1 |
NM_177722.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000397.mm.1 |
TC0100000045.mm.1 |
MTA TC |