Assay Details
| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177722.3 | NP_808390.2 | 4 - 5 | 379 | 95 | |
| GenBank mRNA | CA978963.1 | - | 2 - 3 | 134 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcmdc2
Genomic Map