Assay Details
| Gene Symbol: Gnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008140.2 | NP_032166.1 | 1 - 2 | 152 | 58 | |
| GenBank mRNA | AK149254.1 | - | 1 - 2 | 195 | 58 | |
| BC022793.1 | - | 1 - 2 | 152 | 4505478 | 58 | |
| BC051412.1 | - | 1 - 2 | 174 | 6490986 | 58 | |
| BC058810.1 | - | 1 - 2 | 175 | 6490797 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnat1
Genomic Map