Assay Details
| Gene Symbol: 1700092M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177347.1 | NP_001170818.1 | 1 - 2 | 130 | 62 | |
| GenBank mRNA | AK007043.1 | - | 1 - 2 | 93 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

1700092M07Rik
Genomic Map